Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 19 | 2433 | ||
NM_001162501.1 | NP_001155973.1 | 18 | 4754 | |||
NM_015088.2 | NP_055903.2 | 16 | 4424 | |||
GenBank mRNA | AB029016.2 | - | 16 | 4404 | ||
AK294519.1 | - | 18 | 4745 | |||
AK296590.1 | - | 9 | 1473 | |||
AK304087.1 | - | 13 | 1909 | |||
BC028626.2 | - | 19 | 2366 | 4826787 | ||
BX647630.1 | - | 13 | 3626 |