Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002876.2 | NP_001002876.1 | 5 | 798 | ||
NM_001110215.2 | NP_001103685.1 | 2 | 481 | |||
NM_001304370.1 | NP_001291299.1 | 5 | 1003 | |||
NM_001304372.1 | NP_001291301.1 | 5 | 1088 | |||
NM_001304373.1 | NP_001291302.1 | 4 | 911 | |||
NM_024053.4 | NP_076958.1 | 6 | 890 | |||
GenBank mRNA | AK123479.1 | - | 6 | 1915 | ||
AL160131.1 | - | 6 | 838 | |||
BC000705.2 | - | 6 | 837 | 3349507 | ||
BC007495.2 | - | 6 | 850 | 2966541 | ||
BX332140.2 | - | 4 | 847 | |||
BX417558.2 | - | 4 | 849 | |||
EU035297.1 | - | 2 | 481 |