Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198495.2 | NP_940897.2 | 1 | 1796 | ||
GenBank mRNA | AF338232.1 | - | 1 | 1125 | ||
BE272325.1 | - | 1 | 200 |
Gene Symbol: CTAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278507.1 | NP_001265436.1 | 1 | 2281 | ||
GenBank mRNA | AK292236.1 | - | 1 | 1785 |
Gene Symbol: CTAGE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145659.1 | NP_001139131.1 | 1 | 1747 | ||
GenBank mRNA | BC101322.1 | - | 1 | 1097 |