Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198692.2 | NP_941965.2 | 1 | 1186 | ||
GenBank mRNA | AF086314.1 | - | 1 | 366 | ||
BX103563.1 | - | 1 | 364 |