Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102564.1 | NP_001096034.1 | 6 | 344 | ||
NM_052873.2 | NP_443105.2 | 5 | 359 | |||
NR_045664.1 | 4 | 264 | ||||
NR_045665.1 | 6 | 344 | ||||
GenBank mRNA | AK056735.1 | - | 7 | 412 | ||
AK302944.1 | - | 6 | 327 | |||
BC010436.1 | - | 5 | 330 | 3608220 | ||
BI821833.1 | - | 4 | 240 | |||
CB853974.1 | - | 6 | 305 | |||
DB443802.1 | - | 6 | 343 | |||
DB497730.1 | - | 6 | 327 |