Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 12 | 2205 | ||
NM_023085.3 | NP_075573.2 | 10 | 1740 | |||
GenBank mRNA | AB058748.1 | - | 12 | 2109 | ||
AF089088.1 | - | 12 | 2186 | |||
AF089090.1 | - | 12 | 1881 | |||
AF089091.1 | - | 10 | 1585 | |||
AF089092.1 | - | 11 | 1788 | |||
AF089093.1 | - | 13 | 2100 | |||
AF089094.1 | - | 12 | 2039 | |||
AF089096.1 | - | 4 | 448 | |||
AK074807.1 | - | 12 | 2156 | |||
AK128552.1 | - | 9 | 4002 | |||
BC004260.1 | - | 12 | 2154 | 3606959 | ||
BC007553.1 | - | 12 | 2154 | 2967328 |