Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024044.3 | NP_076949.1 | 3, 4 | 822 | ||
GenBank mRNA | AK027092.1 | - | 3, 4 | 768 | ||
AK225436.1 | - | 3, 4 | 768 | |||
BC000754.2 | - | 3, 4 | 769 | 2823474 | ||
BC000803.1 | - | 3, 4 | 616 | 3452236 | ||
BC015990.2 | - | 3, 4 | 775 | 3503887 | ||
BC019306.1 | - | 3, 4 | 769 | 2823474 |
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014999.2 | NP_001014999.1 | 3, 4 | 822 | ||
GenBank mRNA | CD673028.1 | - | 3, 4 | 607 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 2, 3 | 699 | |||
GenBank mRNA | AK122707.1 | - | 2, 3 | 699 | ||
AK122733.1 | - | 3, 4 | 527 | |||
BC111011.1 | - | 1, 2 | 727 | 5768190 |
Gene Symbol: SLX1B-SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037609.1 | 2, 3 | 699 |