Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015972.3 | NP_057056.1 | 2 | 436 | ||
XM_005266412.1 | XP_005266469.1 | 3 | 1276 | |||
GenBank mRNA | AF077044.1 | - | 2 | 324 | ||
AK000623.1 | - | 2 | 322 | |||
BC000889.1 | - | 2 | 340 | 3462906 | ||
BI253733.1 | - | 2 | 248 |