siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003097.4 | NP_003088.1 | 4 | 475 | ||
NM_022805.3 | NP_073716.1 | 6 | 670 | |||
NM_022806.3 | NP_073717.1 | 6 | 680 | |||
NM_022807.3 | NP_073718.1 | 7 | 815 | |||
NM_022808.3 | NP_073719.1 | 6 | 707 | |||
GenBank mRNA | BC024777.1 | - | 5 | 503 | 3907606 | |
BC025178.1 | - | 4 | 347 | 4158469 | ||
BC035280.1 | - | 8 | 849 |
Gene Symbol: SNURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005678.4 | NP_005669.2 | 4 | 475 | ||
GenBank mRNA | AK098563.1 | - | 4 | 379 | ||
DA134019.1 | - | 4 | 445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




