Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136230.2 | NP_001129702.1 | 6 | 896 | ||
NM_178135.4 | NP_835236.2 | 7 | 1004 | |||
GenBank mRNA | AB073347.1 | - | 7 | 973 | ||
AK292784.1 | - | 7 | 978 | |||
AY186249.1 | - | 7 | 984 | |||
AY186250.1 | - | 6 | 876 | |||
AY268355.1 | - | 7 | 954 | |||
AY358575.1 | - | 7 | 1004 |