Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMZP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012230.3 | NP_036362.3 | 3 | 928 | ||
NM_152992.2 | NP_694537.1 | 3 | 928 | |||
GenBank mRNA | U10099.1 | - | 3 | 874 |
Gene Symbol: POM121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257190.2 | NP_001244119.1 | 6 | 1252 | ||
NM_172020.4 | NP_742017.1 | 4 | 637 | |||
XM_005250726.3 | XP_005250783.1 | 3 | 991 | |||
XM_005250727.3 | XP_005250784.1 | 3 | 991 | |||
XM_006716196.3 | XP_006716259.1 | 6 | 1252 | |||
XM_011516730.1 | XP_011515032.1 | 3 | 991 | |||
XM_017012853.1 | XP_016868342.1 | 6 | 1240 | |||
XM_017012854.1 | XP_016868343.1 | 6 | 880 | |||
GenBank mRNA | AB014518.1 | - | 6 | 1222 | ||
AK123814.1 | - | 4 | 564 | |||
AK292548.1 | - | 3 | 348 | |||
BC008794.2 | - | 3 | 481 | 3621228 |
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099415.2 | NP_001092885.2 | 5 | 1063 | ||
GenBank mRNA | AB354586.1 | - | 3 | 907 | ||
AK293781.1 | - | 1 | 119 | |||
BC082993.1 | - | 5 | 1046 |