Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256570.1 | NP_001243499.1 | 1 | 477 | ||
NM_006917.4 | NP_008848.1 | 1 | 477 | |||
NR_033824.1 | 1 | 477 | ||||
GenBank mRNA | AB307706.1 | - | 1 | 15 | ||
AB593015.1 | - | 1 | 121 | |||
AB593016.1 | - | 1 | 385 | |||
AK312922.1 | - | 1 | 407 | |||
AL538345.3 | - | 1 | 315 | |||
BC012063.1 | - | 1 | 203 | 4635470 | ||
BX118484.1 | - | 1 | 294 | |||
CR456705.1 | - | 1 | 15 | |||
DA251671.1 | - | 1 | 478 | |||
HQ692845.1 | - | 1 | 15 | |||
U38480.1 | - | 1 | 42 |