Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012318.2 | NP_036450.1 | 14 | 3666 | ||
XM_006713884.1 | XP_006713947.1 | 14 | 3371 | |||
XM_017008187.1 | XP_016863676.1 | 13 | 3787 | |||
GenBank mRNA | AF061025.1 | - | 14 | 3666 | ||
BC021208.1 | - | 14 | 3591 | 4126510 |