Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012318.2 | NP_036450.1 | 14 | 5010 | ||
XM_006713884.1 | XP_006713947.1 | 14 | 4715 | |||
XM_017008187.1 | XP_016863676.1 | 13 | 5131 |