Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 7 | 1008 | ||
NM_001008662.2 | NP_001008662.1 | 6 | 908 | |||
GenBank mRNA | AF091090.1 | - | 3 | 210 | ||
AK057176.1 | - | 6 | 724 | |||
AK301326.1 | - | 7 | 695 | |||
AK315917.1 | - | 7 | 705 | |||
AL833593.1 | - | 8 | 810 | |||
AY028624.1 | - | 7 | 828 | |||
BC000819.1 | - | 4 | 381 | 3453829 | ||
CR450282.1 | - | 2 | 139 | |||
CR627392.1 | - | 8 | 827 |