Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198691.2 | NP_941964.2 | 1 | 438 | ||
GenBank mRNA | AB076347.1 | - | 1 | 438 | ||
BC120959.2 | - | 1 | 432 | 40115271 | ||
BC120960.1 | - | 1 | 432 | 40115274 |
Gene Symbol: KRTAP10-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198694.3 | NP_941967.3 | 1 | 386 | ||
GenBank mRNA | AB076352.1 | - | 1 | 386 | ||
BC120949.1 | - | 1 | 385 | 40115143 | ||
BC120950.1 | - | 1 | 385 | 40115146 |