Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024044.3 | NP_076949.1 | 5 | 1006 | ||
NM_178044.2 | NP_835145.1 | 4 | 664 | |||
GenBank mRNA | AK027092.1 | - | 5 | 953 | ||
AK225436.1 | - | 5 | 952 | |||
BC000754.2 | - | 5 | 953 | 2823474 | ||
BC000803.1 | - | 5 | 800 | 3452236 | ||
BC015990.2 | - | 5 | 959 | 3503887 | ||
BC019306.1 | - | 5 | 953 | 2823474 | ||
BC069007.1 | - | 4 | 603 | 4335620 | ||
BC130545.1 | - | 4 | 609 | 40146576 | ||
BC130547.1 | - | 4 | 609 | 40146578 |
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014999.2 | NP_001014999.1 | 5 | 1006 | ||
NM_001015000.2 | NP_001015000.1 | 4 | 664 | |||
GenBank mRNA | BC144462.1 | - | 4 | 609 | 9052988 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 4 | 883 | |||
GenBank mRNA | AK094769.1 | - | 2 | 1365 | ||
AK097386.1 | - | 3 | 529 | |||
AK122707.1 | - | 4 | 884 | |||
AK122733.1 | - | 5 | 711 | |||
BC014471.1 | - | 2 | 148 | 4907290 | ||
BC111011.1 | - | 3 | 911 | 5768190 |
Gene Symbol: SLX1B-SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037609.1 | 4 | 883 |