Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048194.2 | NP_001041659.1 | 11 | 1764 | ||
NM_001048195.2 | NP_001041660.1 | 11 | 1722 | |||
NM_001048199.2 | NP_001041664.1 | 12 | 1846 | |||
NM_001269.4 | NP_001260.1 | 10 | 1671 | |||
NR_030725.1 | 13 | 1989 | ||||
NR_030726.1 | 13 | 1985 | ||||
GenBank mRNA | BC001589.1 | - | 9 | 1608 | ||
BC010067.2 | - | 9 | 1589 | 4122227 | ||
BX109495.1 | - | 1 | 301 | |||
X12654.1 | - | 10 | 1768 |