Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198693.3 | NP_941966.1 | 1 | 962 | ||
NR_130165.1 | 2 | 257 | ||||
GenBank mRNA | AB076348.1 | - | 1 | 962 | ||
AB076349.1 | - | 2 | 257 | |||
AJ566381.1 | - | 1 | 263 |