Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 5 | 1529 | ||
NM_133269.3 | NP_579803.1 | 5 | 1463 | |||
NM_133271.3 | NP_579805.1 | 4 | 1241 | |||
NM_133272.3 | NP_579806.1 | 4 | 1493 | |||
NM_133273.3 | NP_579807.1 | 3 | 1205 | |||
NM_133274.3 | NP_579808.1 | 4 | 1419 | |||
NM_133277.3 | NP_579811.1 | 3 | 1202 | |||
NM_133278.3 | NP_579812.1 | 4 | 1427 | |||
XM_011526625.2 | XP_011524927.1 | 5 | 1453 | |||
XM_017026473.1 | XP_016881962.1 | 5 | 1502 | |||
XM_017026474.1 | XP_016881963.1 | 4 | 1285 |