Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 5 | 1031 | ||
NM_133269.3 | NP_579803.1 | 5 | 965 | |||
NM_133271.3 | NP_579805.1 | 4 | 743 | |||
NM_133272.3 | NP_579806.1 | 4 | 995 | |||
NM_133273.3 | NP_579807.1 | 3 | 707 | |||
NM_133274.3 | NP_579808.1 | 4 | 921 | |||
NM_133277.3 | NP_579811.1 | 3 | 704 | |||
NM_133278.3 | NP_579812.1 | 4 | 929 | |||
XM_011526625.2 | XP_011524927.1 | 5 | 955 | |||
XM_017026473.1 | XP_016881962.1 | 5 | 1004 | |||
XM_017026474.1 | XP_016881963.1 | 4 | 787 | |||
GenBank mRNA | AK290922.1 | - | 5 | 889 | ||
BC027953.1 | - | 5 | 858 | 5211453 | ||
D87853.1 | - | 5 | 768 | |||
D87854.1 | - | 4 | 546 | |||
D87855.1 | - | 4 | 798 | |||
D87856.1 | - | 3 | 510 | |||
D87857.1 | - | 4 | 724 | |||
D87858.1 | - | 5 | 624 | |||
D87859.1 | - | 6 | 846 | |||
D87860.1 | - | 4 | 588 | |||
D87861.1 | - | 3 | 507 | |||
D87862.1 | - | 4 | 732 | |||
DQ075334.1 | - | 5 | 834 | |||
DQ075335.1 | - | 5 | 834 | |||
DQ075336.1 | - | 5 | 834 | |||
DQ075337.1 | - | 5 | 834 | |||
DQ075338.1 | - | 5 | 834 | |||
DQ075339.1 | - | 5 | 834 | |||
S82919.1 | - | 4 | 546 | |||
U43677.1 | - | 4 | 551 | |||
U43774.1 | - | 5 | 773 | |||
X92558.1 | - | 4 | 798 | |||
X92559.1 | - | 4 | 732 | |||
X92560.1 | - | 5 | 768 | |||
X92561.1 | - | 4 | 546 | |||
X92562.1 | - | 3 | 507 |