Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004680.2 | NP_004671.1 | 1 | 196 | ||
NM_170723.1 | NP_733841.1 | 1 | 196 | |||
XM_011531512.1 | XP_011529814.1 | 1 | 198 | |||
GenBank mRNA | AF000981.1 | - | 1 | 151 | ||
AF080597.1 | - | 1 | 151 | |||
AK292375.1 | - | 1 | 218 | |||
BC033041.1 | - | 1 | 201 |
Gene Symbol: CDY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003894.1 | NP_001003894.1 | 1 | 196 | ||
NM_001003895.1 | NP_001003895.1 | 1 | 196 | |||
XM_011531484.1 | XP_011529786.1 | 1 | 198 | |||
GenBank mRNA | AK292318.1 | - | 1 | 169 |