Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003790.2 | NP_003781.1 | 10 | 1398 | ||
NM_148965.1 | NP_683866.1 | 10 | 1425 | |||
NM_148966.1 | NP_683867.1 | 9 | 1287 | |||
NM_148967.1 | NP_683868.1 | 9 | 1263 | |||
NM_148970.1 | NP_683871.1 | 5 | 849 | |||
GenBank mRNA | AF026070.1 | - | 10 | 1405 | ||
AF026071.1 | - | 9 | 1499 | |||
AK094463.1 | - | 4 | 1364 | |||
AY358309.1 | - | 10 | 1412 | |||
BC117189.1 | - | 10 | 1378 | 40125740 | ||
BC143886.1 | - | 9 | 1267 | 9052402 | ||
U74611.1 | - | 10 | 1398 | |||
U75380.1 | - | 10 | 1301 | |||
U83597.1 | - | 10 | 1294 | |||
Y09392.1 | - | 9 | 1479 |