Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177457.4 | NP_803252.1 | 4 | 3368 | ||
NM_177476.3 | NP_803429.1 | 4 | 3387 | |||
NM_177477.3 | NP_803430.1 | 4 | 3501 | |||
GenBank mRNA | AK123335.1 | - | 1 | 1386 | ||
AK126845.1 | - | 1 | 2578 | |||
AL353948.1 | - | 1 | 1028 | |||
BC014560.2 | - | 3 | 2661 | |||
BC047923.1 | - | 1 | 985 |