Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS3 SIRNA Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
UniGene ID:
Hs.494875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s231737
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276260.1
NP_001263189.1
7
1960
NM_001276261.1
NP_001263190.1
15
1884
NM_001276262.1
NP_001263191.1
5
608
NM_001282922.1
NP_001269851.1
8
1818
NM_001282923.1
NP_001269852.1
23
3428
NM_001322215.1
NP_001309144.1
7
1925
NM_130795.3
NP_570613.2
16
2862
NM_134427.2
NP_602299.1
5
882
NM_144488.5
NP_652759.3
26
3766
NM_144489.3
NP_652760.2
5
894
XM_006717219.2
XP_006717282.1
25
3612
XM_006717220.1
XP_006717283.1
23
3388
XM_006717222.1
XP_006717285.1
16
2738
XM_006717225.1
XP_006717288.1
7
1742
XM_006717226.2
XP_006717289.1
8
2035
XM_006717230.3
XP_006717293.1
7
1624
XM_011518894.1
XP_011517196.1
8
1927
XM_011518896.1
XP_011517198.1
8
2289
XM_011518897.2
XP_011517199.1
9
2287
XM_011518898.1
XP_011517200.1
9
2011
XM_011518900.1
XP_011517202.1
8
1812
XM_011518901.1
XP_011517203.1
8
1843
XM_017015003.1
XP_016870492.1
25
3641
XM_017015004.1
XP_016870493.1
22
3136
XM_017015005.1
XP_016870494.1
10
1975
XM_017015006.1
XP_016870495.1
8
2191
GenBank mRNA
AF463495.1
-
16
2862
AF490838.1
-
5
894
AF490839.1
-
23
3240
AF490840.1
-
16
2712
AF493927.1
-
6
1518
AF493941.1
-
6
579
AK054584.1
-
5
871
AK096840.1
-
15
1804
AK123387.1
-
7
1933
AK125059.1
-
23
3399
AK128127.1
-
26
3764
AK226153.1
-
6
3332
AK289666.1
-
8
1654
AY585192.1
-
6
1518
BC018072.1
-
5
685
BC019039.2
-
5
667
BC042555.1
-
16
2767
5175121
U27655.1
-
8
1805
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0