Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282923.1 | NP_001269852.1 | 4 | 418 | ||
NM_017790.4 | NP_060260.3 | 4 | 312 | |||
NM_144488.5 | NP_652759.3 | 7 | 756 | |||
XM_006717219.2 | XP_006717282.1 | 6 | 602 | |||
XM_006717220.1 | XP_006717283.1 | 4 | 378 | |||
XM_011518893.1 | XP_011517195.1 | 7 | 754 | |||
XM_017015003.1 | XP_016870492.1 | 7 | 754 | |||
XM_017015004.1 | XP_016870493.1 | 3 | 130 | |||
GenBank mRNA | AF490839.1 | - | 4 | 233 | ||
AK074977.1 | - | 4 | 282 | |||
AK125059.1 | - | 4 | 389 | |||
AK128127.1 | - | 7 | 754 | |||
DA680346.1 | - | 4 | 312 |