Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323553.1 | NP_001310482.1 | 19, 20 | 2449 | ||
NM_001323554.1 | NP_001310483.1 | 19, 20 | 3388 | |||
NM_001323555.1 | NP_001310484.1 | 19, 20 | 3457 | |||
NM_001323556.1 | NP_001310485.1 | 19, 20 | 2230 | |||
NM_001323557.1 | NP_001310486.1 | 19, 20 | 2299 | |||
NM_001323558.1 | NP_001310487.1 | 20, 21 | 2431 | |||
NM_015137.5 | NP_055952.2 | 19, 20 | 2380 | |||
GenBank mRNA | AK294485.1 | - | 13, 14 | 1519 | ||
AK302799.1 | - | 10, 11 | 1060 | |||
BC047323.1 | - | 3, 4 | 289 | |||
BC071611.1 | - | 19, 20 | 2357 | |||
BC131486.1 | - | 15, 16 | 1715 | |||
BC152442.1 | - | 19, 20 | 2348 | 8862567 | ||
BX648595.1 | - | 6, 7 | 619 | |||
CR749243.1 | - | 19, 20 | 2190 | |||
D63477.1 | - | 19, 20 | 2348 |