Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290814.1 | NP_001277743.1 | 14 | 2712 | ||
GenBank mRNA | BU056443.1 | - | 1 | 20 |