Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015140.3 | NP_055955.1 | 13 | 1815 | ||
GenBank mRNA | AK123789.1 | - | 2 | 884 | ||
AK304464.1 | - | 3 | 301 | |||
AL590888.1 | - | 4 | 548 | |||
BC001070.2 | - | 13 | 1779 | 3504490 | ||
CT841516.1 | - | 13 | 1756 | |||
D63487.1 | - | 13 | 1737 |