Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039555.2 | NP_001034644.2 | 6 | 902 | ||
GenBank mRNA | AK136413.1 | - | 5 | 532 | ||
BC019468.1 | - | 6 | 855 | 4160492 |
Gene Symbol: Cyp2c40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010004.2 | NP_034134.2 | 6 | 934 | ||
GenBank mRNA | AF047727.1 | - | 6 | 848 | ||
BC139471.1 | - | 6 | 934 | 8862493 | ||
BC139472.1 | - | 6 | 934 | 8862494 |