Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111311.1 | NP_001104781.1 | 7 | 884 | ||
NM_001111312.1 | NP_001104782.1 | 8 | 843 | |||
NM_008515.4 | NP_032541.1 | 18 | 1586 | |||
GenBank mRNA | AF045573.1 | - | 18 | 1523 | ||
AK041162.1 | - | 8 | 794 | |||
AK077637.1 | - | 7 | 889 | |||
AK133661.1 | - | 7 | 865 | |||
AK134120.1 | - | 7 | 876 | |||
AK162191.1 | - | 2 | 207 | |||
BC144955.1 | - | 9 | 923 | |||
BC144956.1 | - | 20 | 1544 | 9053504 | ||
BC145642.1 | - | 20 | 1544 | 40130945 |