Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 15 | 5159 | ||
GenBank mRNA | AK086788.1 | - | 1 | 289 | ||
AK122230.1 | - | 11 | 2610 | |||
BC137913.1 | - | 15 | 5165 | 8860932 |