Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026545.3 | NP_080821.3 | 1 | 366 | ||
GenBank mRNA | AK002921.1 | - | 1 | 205 | ||
AK003436.1 | - | 1 | 207 | |||
AK020132.1 | - | 1 | 225 | |||
AK027973.1 | - | 1 | 170 | |||
AK051257.1 | - | 1 | 218 | |||
AK150493.1 | - | 1 | 209 | |||
AK150982.1 | - | 1 | 173 | |||
AK153329.1 | - | 1 | 207 | |||
AK159216.1 | - | 1 | 175 | |||
AK159795.1 | - | 1 | 191 | |||
AK159926.1 | - | 1 | 201 | |||
AK166718.1 | - | 1 | 200 | |||
AK167948.1 | - | 1 | 175 | |||
AK168886.1 | - | 1 | 174 | |||
BC004075.1 | - | 1 | 184 | 3591796 | ||
BC005717.1 | - | 1 | 94 | 2609781 | ||
CF732286.1 | - | 1 | 366 |