Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037726.1 | NP_001032815.1 | 7 | 1015 | ||
NM_009952.2 | NP_034082.1 | 9 | 1179 | |||
NM_133828.2 | NP_598589.2 | 8 | 1137 | |||
GenBank mRNA | AF448507.1 | - | 9 | 1055 | ||
AF448508.1 | - | 1707 | ||||
AK044992.1 | - | 8 | 952 | |||
AK044993.1 | - | 8 | 952 | |||
BC021649.1 | - | 8 | 1070 | 4017170 | ||
M95106.1 | - | 9 | 1089 | |||
U46027.1 | - | 6 | 702 | |||
X92497.1 | - | 7 | 992 |