Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033196.2 | NP_001028368.2 | 11 | 3545 | ||
NM_001291162.1 | NP_001278091.1 | 4 | 480 | |||
GenBank mRNA | AK147564.1 | - | 11 | 3548 | ||
AK147652.1 | - | 11 | 3547 | |||
AK157301.1 | - | 4 | 483 | |||
BC059933.1 | - | 4 | 599 |