Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270996.1 | NP_001257925.1 | 9 | 950 | ||
NM_007404.2 | NP_031430.2 | 9 | 950 | |||
GenBank mRNA | AK122188.1 | - | 9 | 933 | ||
AK148174.1 | - | 9 | 836 | |||
AK155582.1 | - | 9 | 911 | |||
AK155672.1 | - | 9 | 843 | |||
AK161778.1 | - | 9 | 874 | |||
BC047156.1 | - | 9 | 828 | 6488707 | ||
BC052710.1 | - | 9 | 822 | |||
U41765.1 | - | 9 | 822 |