Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013325.4 | NP_037457.3 | 2 | 191 | ||
NM_178326.2 | NP_847896.1 | 2 | 191 | |||
XM_005246992.3 | XP_005247049.2 | 2 | 177 | |||
XM_005246993.2 | XP_005247050.2 | 2 | 177 | |||
XM_005246995.3 | XP_005247052.1 | 2 | 165 | |||
XM_005246996.2 | XP_005247053.1 | 2 | 110 | |||
XM_017003637.1 | XP_016859126.1 | 2 | 290 | |||
XM_017003638.1 | XP_016859127.1 | 2 | 289 | |||
XM_017003639.1 | XP_016859128.1 | 1 | 93 | |||
XM_017003640.1 | XP_016859129.1 | 2 | 99 | |||
GenBank mRNA | AB023160.2 | - | 2 | 98 | ||
AB066215.1 | - | 1 | 88 | |||
AJ504652.1 | - | 1 | 94 | |||
AK027332.1 | - | 1 | 43 | |||
AK027462.1 | - | 2 | 105 | |||
AK027763.1 | - | 2 | 124 | |||
AK125277.1 | - | 2 | 615 | |||
AK296818.1 | - | 2 | 110 | |||
AK298109.1 | - | 2 | 110 | |||
AK302963.1 | - | 3 | 441 | |||
AL080168.1 | - | 2 | 192 | |||
BC000719.2 | - | 2 | 95 | 3504267 | ||
BX648007.1 | - | 2 | 109 |