Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010388.4 | NP_034518.1 | 3 | 577 | ||
GenBank mRNA | AK154923.1 | - | 3 | 681 | ||
BC052864.1 | - | 4 | 832 | |||
BC141105.1 | - | 3 | 643 | 9055717 | ||
BC145385.1 | - | 3 | 643 | 9053943 | ||
BC145386.1 | - | 3 | 643 | 9053944 | ||
BC171960.1 | - | 3 | 643 | 9054246 | ||
KJ650224.1 | - | 3 | 577 | |||
U20314.1 | - | 2 | 523 | |||
U20315.1 | - | 2 | 523 | |||
U20316.1 | - | 2 | 523 | |||
U20317.1 | - | 2 | 523 | |||
U35334.1 | - | 3 | 577 | |||
U35335.1 | - | 3 | 577 | |||
U35336.1 | - | 3 | 577 | |||
U35337.1 | - | 3 | 577 | |||
U35338.1 | - | 3 | 577 | |||
X90796.1 | - | 3 | 577 | |||
X90797.1 | - | 3 | 577 | |||
X90798.1 | - | 3 | 577 | |||
X90799.1 | - | 3 | 577 | |||
X90800.1 | - | 3 | 577 | |||
X90801.1 | - | 3 | 577 | |||
X90802.1 | - | 3 | 577 | |||
X90803.1 | - | 3 | 577 | |||
X90804.1 | - | 3 | 577 | |||
X90805.1 | - | 3 | 577 | |||
X90806.1 | - | 3 | 577 | |||
X90807.1 | - | 3 | 577 | |||
X90808.1 | - | 3 | 577 |
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 3 | 785 | ||
GenBank mRNA | AK032745.1 | - | 3 | 786 | ||
AK169969.1 | - | 3 | 766 | |||
AK170866.1 | - | 3 | 768 | |||
BC002237.1 | - | 747 | 3491702 | |||
BC003718.1 | - | 3 | 725 | 3486844 | ||
KJ650225.1 | - | 3 | 577 | |||
U20309.1 | - | 2 | 523 | |||
U20310.1 | - | 2 | 523 | |||
U20312.1 | - | 2 | 523 | |||
U35329.1 | - | 577 | ||||
U35330.1 | - | 577 | ||||
U35331.1 | - | 577 | ||||
U35332.1 | - | 577 | ||||
U35333.1 | - | 577 | ||||
X62743.1 | - | 685 | ||||
X90964.1 | - | 577 | ||||
X90965.1 | - | 577 | ||||
X90966.1 | - | 577 | ||||
X90967.1 | - | 577 | ||||
X90968.1 | - | 577 | ||||
X90969.1 | - | 577 | ||||
X90970.1 | - | 577 | ||||
X90971.1 | - | 577 | ||||
X90972.1 | - | 577 | ||||
X90973.1 | - | 577 | ||||
X90974.1 | - | 577 | ||||
X90975.1 | - | 577 |