Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198808.1 | NP_001185737.1 | 9, 10 | 1165 | ||
NM_177386.5 | NP_796360.2 | 10, 11 | 1288 | |||
GenBank mRNA | AK052339.1 | - | 9, 10 | 1231 | ||
AK147557.1 | - | 9, 10 | 1170 | |||
AK147651.1 | - | 10, 11 | 1143 | |||
AK220408.1 | - | 7, 8 | 1030 | |||
BC139354.1 | - | 9, 10 | 1076 | 8862376 | ||
BC145610.1 | - | 8, 9 | 1032 | 9054176 |