Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081251.1 | NP_001074720.1 | 19 | 2363 | ||
GenBank mRNA | AK030252.1 | - | 9 | 1154 | ||
AK030781.1 | - | 18 | 2266 | |||
AK165182.1 | - | 18 | 2321 | |||
AK166588.1 | - | 18 | 2317 | |||
BC024438.1 | - | 10 | 1219 | |||
BC029037.1 | - | 17 | 2135 | |||
BC037018.1 | - | 5 | 606 | |||
BC055456.1 | - | 5 | 814 |