Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099628.1 | NP_001093098.1 | 7 | 1202 | ||
GenBank mRNA | AK080580.1 | - | 7 | 1190 | ||
AK085232.1 | - | 7 | 1154 | |||
AK161170.1 | - | 7 | 2724 |