Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110146.1 | NP_001103616.1 | 8 | 704 | ||
NM_021400.3 | NP_067375.3 | 9 | 3098 | |||
GenBank mRNA | AB034730.1 | - | 10 | 2597 | ||
AK132597.1 | - | 3 | 844 | |||
AK149469.1 | - | 8 | 705 | |||
BC130021.1 | - | 7 | 668 |