Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134473.2 | NP_001127945.1 | 12 | 2707 | ||
NM_001278184.1 | NP_001265113.1 | 12 | 2761 | |||
NM_014615.3 | NP_055430.1 | 13 | 2980 | |||
XM_005255859.4 | XP_005255916.3 | 14 | 5122 | |||
XM_005255860.3 | XP_005255917.3 | 13 | 5015 | |||
XM_005255861.4 | XP_005255918.3 | 13 | 4903 | |||
XM_005255863.4 | XP_005255920.1 | 13 | 2872 | |||
XM_005255864.4 | XP_005255921.1 | 13 | 3604 | |||
XM_005255865.4 | XP_005255922.1 | 12 | 3385 | |||
XM_011522965.2 | XP_011521267.1 | 13 | 3371 | |||
XM_011522966.2 | XP_011521268.1 | 13 | 2838 | |||
XM_017023083.1 | XP_016878572.1 | 13 | 3130 | |||
GenBank mRNA | AK127344.1 | - | 2 | 1391 | ||
AK308612.1 | - | 3 | 429 | |||
BC037556.1 | - | 12 | 2730 | 4994198 | ||
D80004.2 | - | 12 | 2645 |