Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252218.1 | NP_001239147.1 | 4 | 826 | ||
NM_001252219.1 | NP_001239148.1 | 5 | 684 | |||
NM_053257.3 | NP_444487.1 | 5 | 612 | |||
GenBank mRNA | AK014295.1 | - | 5 | 545 | ||
BC008223.1 | - | 4 | 534 |