Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam208b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134063.3 | NP_598824.3 | 16 | 7263 | ||
GenBank mRNA | AK128934.1 | - | 3 | 2580 | ||
AK131707.1 | - | 5 | 4659 | |||
AK220437.1 | - | 16 | 7251 | |||
BC016423.1 | - | 3 | 1947 | |||
BC045617.1 | - | 3 | 2286 | |||
BC085160.1 | - | 7 | 6006 |