Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054051.1 | NP_473392.1 | 4 | 590 | ||
GenBank mRNA | AK089414.1 | - | 4 | 580 | ||
AK146559.1 | - | 4 | 640 | |||
AK170392.1 | - | 4 | 517 | |||
AK171497.1 | - | 4 | 580 | |||
AY050219.1 | - | 4 | 419 | |||
BC047282.1 | - | 4 | 590 | 6491155 |