Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016748.2 | NP_058028.2 | 9, 10 | 1148 | ||
GenBank mRNA | AK146476.1 | - | 9, 10 | 1149 | ||
AK148259.1 | - | 9, 10 | 1097 | |||
AK161110.1 | - | 9, 10 | 1094 | |||
BC006698.1 | - | 9, 10 | 1123 | 3498717 | ||
U49350.1 | - | 9, 10 | 1121 |