Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122960.1 | NP_001116432.1 | 3 | 350 | ||
NM_026070.3 | NP_080346.1 | 3 | 350 | |||
NM_027487.1 | NP_081763.1 | 3 | 350 | |||
GenBank mRNA | AK004206.1 | - | 3 | 265 | ||
AK009358.1 | - | 3 | 246 | |||
AK013835.1 | - | 3 | 355 | |||
AK017732.1 | - | 3 | 278 | |||
AK028599.1 | - | 2 | 783 | |||
AK158822.1 | - | 3 | 351 | |||
BC029256.1 | - | 3 | 247 | 3661472 | ||
CK795584.1 | - | 3 | 302 |