Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270940.1 | NP_001257869.1 | 8 | 1682 | ||
NM_015171.3 | NP_055986.1 | 7 | 1546 | |||
XM_005255197.3 | XP_005255254.1 | 5 | 880 | |||
XM_005255198.1 | XP_005255255.1 | 3 | 1029 | |||
XM_005255199.2 | XP_005255256.1 | 1 | 158 | |||
XM_011545772.2 | XP_011544074.1 | 7 | 1215 | |||
GenBank mRNA | AB002368.3 | - | 7 | 1497 | ||
AK026172.1 | - | 4 | 690 | |||
AK123846.1 | - | 8 | 1314 | |||
AY026388.1 | - | 7 | 1019 | |||
BC004403.1 | - | 1 | 286 | |||
BC014071.2 | - | 1 | 286 | |||
BC078674.1 | - | 3 | 491 | |||
BC108286.1 | - | 3 | 448 | |||
BC130304.1 | - | 7 | 1019 | 40146335 | ||
BC144189.1 | - | 7 | 1019 | 9052709 |