Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081314.2 | NP_001074783.1 | 2 | 1234 | ||
GenBank mRNA | AK014341.2 | - | 1 | 817 | ||
BC039571.1 | - | 2 | 1233 | 1515262 |